VWR®, Micro cover glasses, rectangular
24×50 mm
VWRU48393-241
These micro cover glasses are made of glass, and each set of numbered slides is uniform in size and thickness.
The No. 1 slides are 0,13 to 0,17 mm in thickness. The No. 1.5 slides are 0,16 to 0,19 mm in thickness.
- 길이×너비:24×50 mm
- Seq:15