VWR®, 원심분리 튜브
15, Screw cap, 3000×g, sterile, 25/rack, 20 packs/case
공급사: VWR Collection
Jump to:
Product Details & Documents
PS, 투명, 원뿔형, 일체형 밀봉이 있는 플랫 캡 포함
- Electron beam irradiated
- Longer length screw caps with sealing ring prevent any leakage
- Engraved graduations at the conical bottom of each tube
- Graduations and writing area are chloroform-resistant
- During the freezing process, it is better to use plastic rack
- During high-speed centrifugation, it is better to use suitable rotor adapters
사양
- Closure type:Screw cap
- Packed:25/rack, 20 packs/case
- Version:sterile
- 용량:15
- Seq:02
- RCF max.:3000×g
- Cat. No.:525-0639
- Environmental certification:ISO 14001/Certificate No.: 04923E00243R0M
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



