VWR®, N-Plan Objectives
IOS N-plan achromatic objective, 20×/0,40 mm, 1,5 mm
제품 번호 630-1641
공급사: VWR Collection
Jump to:
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
VWR® for Microscopy
Product Literature
사양
- 확대:20×/0,40 mm
- 설명:IOS N-plan achromatic objective
- Seq:203
- Working distance:1,5 mm