VWR®, N-Plan Objectives
IOS N-plan achromatic objective, 40×/0,65 mm, 0,45 mm
제품 번호 630-1642
공급사: VWR Collection
Jump to:
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
VWR® for Microscopy
제품 브로셔
사양
- 확대:40×/0,65 mm
- 설명:IOS N-plan achromatic objective
- Seq:204
- Working distance:0,45 mm