Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
₩0
주문 입력
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Korea, Republic of
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
깔때기
Separating Funnels
분리 깔때기, conical
분리 깔때기, conical
Print
분리 깔때기, conical
2000, 29/32 NS
Catalog # 532-0017
Certificates
Supplier:
VWR Collection
분리 깔때기, conical
Catalog # 532-0017
Supplier:
VWR Collection
Certificates
Expand image
Return to Top
분리 깔때기, conical
2000, 29/32 NS
Specifications
Add to cart
Jump to:
Specifications
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Inquire For Price
Other Product Family Options Available
Return Policy
Product Details & Documents
NS-PE 마개가 있는 붕규산염 3.3 유리.
눈금 포함/미포함
PTFE 키가 있는 스톱콕
매우 우수한 내화학성 및 온도 저항성
Documents
CoC VWR Separating Funnels Conical with NS-PE Stopper
coc_VWRI532-0006_GL0825
coc_VWRI532-0006_GL2522
coc_VWRI532-0007_GL1722
coc_VWRI532-0007_GL1925
More
Product Literature
VWR® Glassware Solutions
Specifications
Socket size:
29/32 NS
용량:
2000
Seq:
12