₩0
주문 입력
/
Products
Services
Digital Solutions
Applications
Support
About
Korea, Republic of
Orders Link
ContactUsLinkComponent
깔때기
Separating Funnels
분리 깔때기, conical
분리 깔때기, conical
100, 19/26 NS
532-0007
:
VWR Collection
분리 깔때기, conical
532-0007
:
VWR Collection
분리 깔때기, conical
100, 19/26 NS
1 * 1 items
-
+
NS-PE 마개가 있는 붕규산염 3.3 유리.
눈금 포함/미포함
PTFE 키가 있는 스톱콕
매우 우수한 내화학성 및 온도 저항성
CoC VWR Separating Funnels Conical with NS-PE Stopper
coc_VWRI532-0006_GL0825
coc_VWRI532-0006_GL2522
coc_VWRI532-0007_GL1722
coc_VWRI532-0007_GL1925
VWR® Glassware Solutions
Socket size:
19/26 NS
용량:
100
Seq:
02
VWR®, Filtering Flasks, with Glass Hose Connection
#: 511-0261
: VWR Collection
VWR®, Glass Stoppers for Standard Ground Joints
#: 217-0889
: VWR Collection
VWR® Flasks, Round Bottom, with Standard Ground joint
#: 201-1354
: VWR Collection