VWR® Redox Electrodes
RD 223 (redox), BNC, 1 m cable, Gel, Glass, 12×120 mm
제품 번호 662-1409
공급사: VWR Collection
Jump to:
Product Details & Documents
사양
- Connection:BNC, 1 m cable
- 직경×길이:12×120 mm
- 설명:RD 223 (redox)
- Material:Glass
- Electrolyte:Gel