VWR®, Bouffant Caps, PP, Detectable
Blue, 610 mm
제품 번호 113-1579
공급사: VWR Collection
Jump to:
Product Details & Documents
These detectable caps, made of non woven polypropylene, provide an effective way to cover the hair when working in hygienic environments.
- Increased detectability thanks to foil strip
- Two elastic bands for improved fit
Weight: 10 g/m².
사양
- 직경:610 mm
- Colour:Blue
- Cat. No.:113-1579
- Garment application:Laboratory