VWR®, Accessories for Circulators
PT100 probe with 15 m cable
제품 번호 462-0274
공급사: VWR Collection
Jump to:
Product Details & Documents
Stainless steel probes suitable for used with any instrument that has an Advanced Digital (AD) or Advanced Programmable (AP) controller.
Probe measurements: 152 mm length, 4,7 mm Ø.
Documents
사양
- 설명:PT100 probe with 15 m cable