VWR®, Accessories for Circulators
PT100 probe with 3 m cable
Catalog # 462-0272
Supplier: VWR Collection
Jump to:
Product Details & Documents
Stainless steel probes suitable for used with any instrument that has an Advanced Digital (AD) or Advanced Programmable (AP) controller.
Probe measurements: 152 mm length, 4,7 mm Ø.
Documents
VWR® for baths, thermostats and circulators
Product Literature
Specifications
- 설명:PT100 probe with 3 m cable



