VWR®, Accessories for Circulators
PT100 probe with 0,6 m cable
462-0271
Stainless steel probes suitable for used with any instrument that has an Advanced Digital (AD) or Advanced Programmable (AP) controller.
Probe measurements: 152 mm length, 4,7 mm Ø.
- 설명:PT100 probe with 0,6 m cable