주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
일회용 봉투

일회용 봉투

Autoclavable bags, clear, 16, 780×400 mm
Catalog # 129-0581
Supplier:   VWR Collection
일회용 봉투
Catalog # 129-0581
Supplier:  VWR Collection
일회용 봉투
일회용 봉투

Autoclavable bags, clear, 16, 780×400 mm

Price
Inquire For Price
Log in for availability
1 * 500 items
Product Details & Documents

피펫 팁, 일회용 페트리 디쉬, 샘플 튜브 같은 실험실 폐기물의 안전한 처리.
PP, 40 µm 두께, 투명, 'Biohazard' 문구 유무.

  • 최대 145°C의 내열성
  • 오토클레이브 가능
Specifications
  • 용량:
    16
  • 설명:
    Autoclavable bags, clear
  • 길이×너비:
    780×400 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.