일회용 봉투
Autoclavable bags, clear, 12, 660×305 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
피펫 팁, 일회용 페트리 디쉬, 샘플 튜브 같은 실험실 폐기물의 안전한 처리.
PP, 40 µm 두께, 투명, 'Biohazard' 문구 유무.
- 최대 145°C의 내열성
- 오토클레이브 가능
Specifications
- 용량:12
- 설명:Autoclavable bags, clear
- 길이×너비:660×305 mm
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



