VWR® Talon®, Stand plate, rectangular
Support stand base, rectangular, 152×229 mm
제품 번호 VWRU12985-072
공급사: VWR Collection
Jump to:
Product Details & Documents
Cast iron with black enamel finish.
- All bases feature a built-in support rod holder with locking knob that accepts a 13 mm Ø support rod (sold separately)
- For use with VWR® Talon® lab frames
사양
- 설명:Support stand base, rectangular
- 길이×너비:152×229 mm
- Seq:03