Safranine O solution
1 L
공급사: VWR Chemicals
Jump to:
Product Details & Documents
동의어: CI 50240
- CAS Number:477-73-6
- 화학식:C₂₀H₁₉ClN₄
- MDL 번호:MFCD00011759
- Molecular Weight:350.85 g/mol
- EINECS:207-518-8
- UN:0000
Documents
사양
사양
- Pk:1 L
사양
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



