주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Micro cover glasses, rectangular

VWR®, Micro cover glasses, rectangular

22×50 mm
제품 번호 VWRU48393-059
공급사:   VWR Collection
VWR®, Micro cover glasses, rectangular
제품 번호 VWRU48393-059
공급사:  VWR Collection
VWR®, Micro cover glasses, rectangular
VWR®, Micro cover glasses, rectangular

22×50 mm

Jump to:
Price
₩23,818
Log in 재고 확인
1 * 1 items
Product Details & Documents

These micro cover glasses are made of glass, and each set of numbered slides is uniform in size and thickness.

The No. 1 slides are 0,13 to 0,17 mm in thickness. The No. 1.5 slides are 0,16 to 0,19 mm in thickness.

사양
  • 길이×너비:
    22×50 mm
  • Seq:
    03