Orders LinkContactUsLinkComponent
VWR®, Micro Spatulas
VWR®, Micro Spatulas
165 mm
제품 번호 VWRU82027-520
공급사:   VWR Collection
VWR®, Micro Spatulas
제품 번호 VWRU82027-520
공급사:  VWR Collection
VWR®, Micro Spatulas
VWR®, Micro Spatulas
165 mm
Jump to:
Price
₩20,142
Log in 재고 확인
1 * 1 items
Product Details & Documents
Stainless steel. These micro spatulas are ideal for small quantity applications such as sample transfer into test tubes or microcentrifuge tubes.
  • Rustproof material
  • With non stick surfaces
  • Mirror finish
They feature one rounded end and one tapered end, allowing for a variety of transfer, mixing, or handling applications.
사양
  • 길이:
    165 mm
  • Seq:
    01