Orders LinkContactUsLinkComponent
VWR®, Dissecting Scissors
VWR®, Dissecting Scissors
Straight, sharp/sharp, 165 mm
제품 번호 VWRU82027-592
공급사:   VWR Collection
VWR®, Dissecting Scissors
제품 번호 VWRU82027-592
공급사:  VWR Collection
VWR®, Dissecting Scissors
VWR®, Dissecting Scissors
Straight, sharp/sharp, 165 mm
Jump to:
Price
₩34,418
Log in 재고 확인
1 * 1 items
Product Details & Documents
Stainless steel.
사양
  • Type:
    Straight, sharp/sharp
  • 길이:
    165 mm
  • Seq:
    07