VWR® Basic, Bouffant Caps, Latex-Free
White, 530 mm
제품 번호 VWRU89107-768
공급사: VWR Collection
Jump to:
Product Details & Documents
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Available in a wide range of colours
Weight: 10 g/m²
: 100 pieces per bag, 10 bags per case.
사양
- 직경:530 mm
- Colour:White
- Seq:17



