정량 플라스크
2000, 29/32, 0,600
공급사: VWR Collection
Jump to:
Product Details & Documents
붕규산염 3.3 유리, 클래스 A.
- PP white stopper
- Marks and inscriptions in blue enamel
- Calibrated to contain (TC, In)
DIN EN ISO 1042
Certifications: DIN EN ISO 1042
Ordering information: 배치 넘버와 인증서가 포함됩니다.
Documents
CoC VWR Volumetric Flask, Class A, NS Plastic Stopper.pdf COC_VWRI612-3738_03.19 COC_VWRI612-3738_GL0119 COC_VWRI612-3738_GL0122 coc_VWRI612-3738_GL0325
More
VWRI612-3738_TDS_EN VWRI612-3740_TDS_EN VWRI612-3744_TDS_EN VWRI612-3745_TDS_EN VWRI612-3820_TDS_EN
Datasheet
VWR® for Calibrations VWR® Glassware Solutions
Product Literature
사양
- NS:29/32
- 용량:2000
- Seq:11
- Tolerance:0,600