정량 플라스크
200, 14/23, 0,150
붕규산염 3.3 유리, 클래스 A.
- PP white stopper
- Marks and inscriptions in blue enamel
- Calibrated to contain (TC, In)
DIN EN ISO 1042
Certifications: DIN EN ISO 1042
Ordering information: 배치 넘버와 인증서가 포함됩니다.
Documents
CoC VWR Volumetric Flask, Class A, NS Plastic Stopper.pdf COC_VWRI612-3738_03.19 COC_VWRI612-3738_GL0119 COC_VWRI612-3738_GL0122 coc_VWRI612-3738_GL0325
VWRI612-3738_TDS_EN VWRI612-3740_TDS_EN VWRI612-3744_TDS_EN VWRI612-3745_TDS_EN VWRI612-3820_TDS_EN
Datasheet
VWR® for Calibrations VWR® Glassware Solutions
Product Literature
- NS:14/23
- 용량:200
- Seq:07
- Tolerance:0,150