VWR®, Micro Spatulas
203 mm
제품 번호 VWRU82027-518
공급사: VWR Collection
Jump to:
Product Details & Documents
Stainless steel. These micro spatulas are ideal for small quantity applications such as sample transfer into test tubes or microcentrifuge tubes.
- Rustproof material
- With non stick surfaces
- Mirror finish
They feature one rounded end and one tapered end, allowing for a variety of transfer, mixing, or handling applications.
사양
- 길이:203 mm
- Seq:04