VWR®, Double Spatulas
Round/tapered end, 178 mm
제품 번호 VWRU82027-490
공급사: VWR Collection
Jump to:
Product Details & Documents
These stainless steel spatulas feature one rounded end and one tapered end.
- Rustproof material
- Substances will not stick to the surfaces
Ideal for the transfer or collection of small sample amounts.
사양
- 설명:Round/tapered end
- 길이:178 mm
- Seq:02