VWR® Basic Beard Covers, SPP
With latex, White
113-1463
VWR® Basic SPP beard covers are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility, lot-controlled and validated through independent laboratory testing.
Diameter (stretched): 406 mm
- Colour:White
- 설명:With latex
- Seq:01