VWR® Basic Frocks, SPP
White, XL
공급사: VWR Collection
Jump to:
Product Details & Documents
VWR® Basic SPP frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Snap collar, raglan sleeves and snap closure
- Elasticated cuffs, without pockets
- Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility that is lot controlled and validated through independent laboratory testing.
사양
- 의류 사이즈:XL
- Colour:White
- Cat. No.:113-1153
- Garment application:Cleanroom|Laboratory
- Gender:Unisex
- Pockets:Without pockets
- Closure type:Press studs / snaps
- Weight:48 g/m²