Orders LinkContactUsLinkComponent
클린룸 작업복, VWR® Maximum, SF
클린룸 작업복, VWR® Maximum, SF
White, XXL
제품 번호 113-1077
공급사:   VWR Collection
클린룸 작업복, VWR® Maximum, SF
제품 번호 113-1077
공급사:  VWR Collection
클린룸 작업복, VWR® Maximum, SF
클린룸 작업복, VWR® Maximum, SF
White, XXL
Jump to:
Price
가격 문의
Log in 재고 확인
1 * 25 items
Product Details & Documents
VWR® Maximum SF overalls are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
  • Available as a standard overall with collar, either elasticated ankles or non slip boot covers, or hooded overall with fluid-impervious boot covers
  • Inset sleeves, zip closure, elasticated cuffs
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding: Level 1 Helmke drum classification
  • Light chemical splash resistance
  • Improved breathability to ensure user comfort
  • Soft, cloth-like fabric with maximum tear strength
Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
사양
  • 의류 사이즈:
    XXL
  • Colour:
    White
  • Seq:
    20
  • Cat. No.:
    113-1077
  • Model:
    SF
  • Garment application:
    Cleanroom|Laboratory
  • Bacterial filtration efficiency:
    99,9%