VWR® Basic Cleanroom Overalls, SPP
Without hood, White, L
공급사: VWR Collection
Jump to:
Product Details & Documents
VWR® Basic SPP overalls are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Tapered collar, inset sleeves, zip closure, elasticated cuffs
- Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms.
Tested and manufactured in an ISO certified facility also lot-controlled and validated through independent laboratory testing.
사양
- 의류 사이즈:L
- Colour:White
- 설명:Without hood
- Cat. No.:113-1105
- Garment application:Cleanroom|General Purpose|Laboratory