ContactUsLinkComponent
Deepwell plate 24 wells, 10ml, virgin PP Max. volume: 10,0 ml/well Working volume: 50 µl to 9,9 ml V-shaped well bottom 1 * 20 items
제품 번호 RITT43001-0066
공급사:  RITTER
undefined
Deepwell plate 24 wells, 10ml, virgin PP Max. volume: 10,0 ml/well Working volume: 50 µl to 9,9 ml V-shaped well bottom 1 * 20 items
제품 번호 RITT43001-0066
공급사:  RITTER
관련 정보
Frequently Bought Together