Deepwell plate 48 wells, 5ml, PP Max. volume: 5,0 ml/well Working volume: 50 µl to 4,9 ml Optimized retained storage volume due to rectangular well shape V-shaped well bottom 1 * 20 items
제품 번호 RITT43001-0062
공급사: RITTER
Frequently Bought Together