VWR®, Sample library tubes
Individual tubes, Clear
VWRU82006-698
PP, clear.
- Autoclavable and freezable (–80 °C)
- Printed with an alpha numeric grid that helps to find a sample easily
These 1,2 ml sample library tubes are supplied in a 96 tube format for storage of specimens. The rack is designed to meet SBS standards and can be used in robotic applications.
: Supplied in 8×12 racks with clear, snug fit lid.
- Colour:Clear
- 설명:Individual tubes
- Seq:03