VWR®, Specimen forceps, serrated tips
Straight, serrated tips, 305 mm
제품 번호 VWRU82027-432
공급사: VWR Collection
Jump to:
Product Details & Documents
These specimen forceps made from stainless steel are ergonomically designed for safely and efficiently removing specimens from deep jars or bottles.
- Serrated tips
- Easy to use
사양
- Type:Straight, serrated tips
- 길이:305 mm
- Seq:08