주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
3중 디스플레이 온도계, Traceable

3중 디스플레이 온도계, Traceable

−50…+70 °C, ±1,0, 1,0 °C, Bottle-buffered sensor, 70×20×120 mm, 113 g
제품 번호 620-1661
공급사:   VWR Collection
3중 디스플레이 온도계, Traceable
제품 번호 620-1661
공급사:  VWR Collection
3중 디스플레이 온도계, Traceable
3중 디스플레이 온도계, Traceable

−50…+70 °C, ±1,0, 1,0 °C, Bottle-buffered sensor, 70×20×120 mm, 113 g

Jump to:
Price
가격 문의
Log in 재고 확인
1 * 1 items
Product Details & Documents

These digital thermometers are suitable for monitoring of temperatures in freezers, water baths, heating blocks, incubators and refrigerators. The bottle-enclosed, temperature-buffered sensor is insulated from transient temperature changes. Micro cable allows refrigerator doors to close on it.

  • Maximum/minimum monitors high and low readings overnight, at the weekend or for any time period
  • Programmable alarm sounds continuously when temperature rises above/falls below set points and can be turned off manually
  • Visual and audible signals continue even if temperature returns to non alarm range

Waterproof probe (Lר: 19×8 mm) or bottle-buffered sensor, with 3 metre cable.

Conforms to ISO 17025 and is traceable to NIST.

Patented unit fullfils all CDC thermometer and vaccine thermometer requirements.

Certifications: Compliance with ISO/IEC 17025, ANSI/NCSL Z541-1 and 9001. Traceability to NIST. Patented unit fulfils all CDC thermometer and vaccine thermometer requirements.

납품 정보: Supplied with Traceable® certificate of calibration, flip stand, wall bracket, VELCRO mounting strips and batteries.

사양
  • Range:
    −50…+70 °C
  • 해상도:
    1,0 °C
  • Weight:
    113 g
  • Seq:
    03
  • W×D×H:
    70×20×120 mm
  • Accuracy:
    ±1,0
  • Probe:
    Bottle-buffered sensor
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.