VWR® Talon®, Stand plate, rectangular
Support stand base, rectangular, 203×254 mm
Catalog # VWRU12985-074
Supplier: VWR Collection
Jump to:
Product Details & Documents
Cast iron with black enamel finish.
- All bases feature a built-in support rod holder with locking knob that accepts a 13 mm Ø support rod (sold separately)
- For use with VWR® Talon® lab frames
Specifications
- 설명:Support stand base, rectangular
- 길이×너비:203×254 mm
- Seq:04