VWR®, Spatulas
Standard blade, 304 mm, 152
제품 번호 VWRU82027-462
공급사: VWR Collection
Jump to:
Product Details & Documents
Stainless steel spatulas with wooden handle. Economical tools for handling, transferring, blending, or collecting chemical and biological samples.
- Sturdy and durable
- Stainless steel construction
- Multiple sizes allow for varied laboratory applications
사양
- 설명:Standard blade
- 길이:304 mm
- Seq:04
- Blade length:152