VWR®, Dissecting forceps
Straight, serrated, medium pointed tips, 114 mm
Catalog # VWRU82027-398
Supplier: VWR Collection
Jump to:
Product Details & Documents
Dissecting forceps made of stainless steel. Available straight or curved and in different lengths.
Specifications
- Type:Straight, serrated, medium pointed tips
- 길이:114 mm
- Seq:06