주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Micro cover glasses, rectangular

VWR®, Micro cover glasses, rectangular

24×50 mm
제품 번호 VWRU48393-081
공급사:   VWR Collection
VWR®, Micro cover glasses, rectangular
제품 번호 VWRU48393-081
공급사:  VWR Collection
VWR®, Micro cover glasses, rectangular
VWR®, Micro cover glasses, rectangular

24×50 mm

Jump to:
Price
₩22,097
Log in 재고 확인
1 * 1 oz
Product Details & Documents

These micro cover glasses are made of glass, and each set of numbered slides is uniform in size and thickness.

The No. 1 slides are 0,13 to 0,17 mm in thickness. The No. 1.5 slides are 0,16 to 0,19 mm in thickness.

사양
  • 길이×너비:
    24×50 mm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.