VWR®, Electrode cleaning solutions
Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins, Plastic bottle, 100 mL
For full information concerning health and safety data please see the website.
- 포장 유형:Plastic bottle
- Pk:100 mL
- 설명:Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins