VWR®, pH Electrodes
pH electrode, micro, Non refillable, micro, 0 - 14, 0...60, Epoxy, Mini DIN
제품 번호 662-1814
공급사: VWR Collection
Jump to:
사양
- Connection:Mini DIN
- Type:Non refillable, micro
- 설명:pH electrode, micro
- Material:Epoxy
- Temp.:0...60
- pH:0 - 14