주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
 

VWR®, Polyester Films with Colour-Coded Marking Areas

Films with red dot, Transparent, 77,5 µm, -
제품 번호 391-1269
공급사:   VWR Collection
VWR®, Polyester Films with Colour-Coded Marking Areas
제품 번호 391-1269
공급사:  VWR Collection
VWR®, Polyester Films with Colour-Coded Marking Areas
VWR®, Polyester Films with Colour-Coded Marking Areas

Films with red dot, Transparent, 77,5 µm, -

Jump to:
Price
가격 문의
Log in 재고 확인
1 * 100 items
Product Details & Documents

Identical to polyester films above, but include a colour dot and white writing area on each end tab for plate identification. End tabs with backing removed adhere to the ends of the plate and are perforated so they can be left in place even if the centre portion of the film is removed. Alternatively, one inscribed colour-coded end tab can be removed and pasted in a laboratory notebook while the other remains on the plate.

  • Choice of five colours for easy plate identification
  • Applications: Short-term storage, incubation and ELISA
  • Recommended temperature range −40...+120 °C
사양
  • 광학 특성:
    Transparent
  • Sterile:
    -
  • 설명:
    Films with red dot
  • 두께:
    77,5 µm
  • Cat. No.:
    391-1269