₩0
주문 입력
/
Products
Services
Digital Solutions
Applications
Support
About
Korea, Republic of
Orders Link
ContactUsLinkComponent
Forceps
VWR® Forceps
VWR® Forceps
Sharp end, 100 mm
232-0151
:
VWR Collection
VWR® Forceps
232-0151
:
VWR Collection
VWR® Forceps
Sharp end, 100 mm
1 * 1 items
-
+
PTFE, white.
Very good chemical resistance, inert, non stick
Temperature resistant from –200 to +280 °C
Smooth surfaces prevent contamination and allow easy cleaning
232-0150
232-0151
CoC_VWR_FORCEPS_WHITE
VWRI232_0191_20240301.pdf
Type:
Sharp end
길이:
100 mm