VWR® Forceps
Square end, 100 mm
공급사: VWR Collection
Jump to:
Product Details & Documents
PTFE, white.
- Very good chemical resistance, inert, non stick
- Temperature resistant from –200 to +280 °C
- Smooth surfaces prevent contamination and allow easy cleaning
Documents
사양
- Type:Square end
- 길이:100 mm