VWR®, Dropping Bottles, PTFE
50, 100 mm, 43 mm
제품 번호 216-0457
공급사: VWR Collection
Jump to:
Product Details & Documents
PTFE, white.
- Flexible wall for easy use
- Screw-fit cap
- Guaranteed chemically resistant and inert
Suitable for dropping-out aggressive chemicals.
Documents
사양
- 외부 직경:43 mm
- 용량:50
- 높이:100 mm