VWR®, Calibration Weights, Class 1
1000 g, 2500
제품 번호 611-8763
공급사: VWR Collection
Jump to:
Product Details & Documents
Stainless steel masses adjusted to class 1 for precise checking and adjustment of electronic balances.
Certifications: ASTM certified.
납품 정보: Individual weights are supplied without certificate.
Documents
사양
- Weight:1000 g
- Seq:05
- Tolerance:2500