VWR®, 일회용 마이크로 계량 접시
0,11, 10×4×4,5 mm
제품 번호 611-1358
공급사: VWR Collection
Jump to:
Product Details & Documents
Aluminium.
These micro weighing boats are compatible with most commercially available analysers and microbalances, and are packed in easy to use plastic containers for storage and dispensing.
사양
- 용량:0,11
- Seq:06
- W×D×H:10×4×4,5 mm