VWR®, 일회용 마이크로 계량 접시
0,35, 13×3,5 mm
611-1356
Aluminium.
These micro weighing boats are compatible with most commercially available analysers and microbalances, and are packed in easy to use plastic containers for storage and dispensing.
- Int.Ø×H:13×3,5 mm
- 용량:0,35
- Seq:03