VWR®, 일회용 마이크로 계량 접시
0,035, 6×2,5 mm
제품 번호 611-1355
공급사: VWR Collection
Jump to:
Product Details & Documents
Aluminium.
These micro weighing boats are compatible with most commercially available analysers and microbalances, and are packed in easy to use plastic containers for storage and dispensing.
사양
- Int.Ø×H:6×2,5 mm
- 용량:0,035
- Seq:01