주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
LB 배지, pre-mixed

LB 배지, pre-mixed

LB broth, Miller formulation, tissue culture grade, 2 kg
Catalog # J106-2KG
Supplier:   VWR Chemicals
LB 배지, pre-mixed
Catalog # J106-2KG
Supplier:  VWR Chemicals
CAS Number:  
LB 배지, pre-mixed
LB 배지, pre-mixed

LB broth, Miller formulation, tissue culture grade, 2 kg

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 2 kg
Product Details & Documents

Lysogeny broth (LB)는 E. coli.의 번식과 유지에 사용되는 영양이 풍부한 배지입니다. 또한 플라스미드 DNA 및 재조합 단백질의 제조에도 널리 사용됩니다.

  • 사전 혼합 배지를 사용하여 시간 절약
  • 균일한 고품질 파우더

Formulation of 10 g tryptone, 10 g NaCl, and 5 g yeast extract (25 g powder total) per L of liquid broth.
The instructions included on the label state to prepare LB broth by dissolving 25 g powder per liter in deionized water.

Delivery information: 개별 ReadyPack™ 파우치는 1L의 배지를 준비합니다.

Specifications

Specifications

  • Pk:
    2 kg
  • 설명:
    LB broth, Miller formulation, tissue culture grade

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.