주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
 

Silver sulphate ≥98%, TECHNICAL

Plastic bottle for solids, 50 g
  21589.153
 
Silver sulphate ≥98%, TECHNICAL
  21589.153
 :  10294-26-5
Silver sulphate ≥98%, TECHNICAL
Silver sulphate ≥98%, TECHNICAL

Plastic bottle for solids, 50 g

 :  10294-26-5
 
₩631,964
   
1 * 50 g
 
 :  Disilver sulphate
  •  :10294-26-5
  •  :Ag₂SO₄
  •  :MFCD00003407
  •  :311.8 g/mol
  •  :13,08603
  •  :1085 °C (1013 hPa)
  •  :655 °C
  •  :5,45 g/cm³ (20 °C)
  •  :Ambient
  •  :233-653-7
  •  :3077
  •  :9,III
 

 

  • Assay:
    Min. 98 %

 

  • 포장 유형:
    Plastic bottle for solids
  • Pk:
    50 g