VWR®, Calibration Weights, Class M2
100 g, 16
제품 번호 611-8376
공급사: VWR Collection
Jump to:
Product Details & Documents
Test weights in either stainless steel or aluminium.
납품 정보: Supplied in a plastic box.
사양
- Weight:100 g
- Seq:10
- Tolerance:16
Test weights in either stainless steel or aluminium.
납품 정보: Supplied in a plastic box.