VWR®, Calibration Weights, Class M1
50 g, 3,0
Catalog # 611-8286
Supplier: VWR Collection
Jump to:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Delivery information: All weights are supplied in plastic cases.
Documents
Specifications
- Weight:50 g
- Seq:15
- Tolerance:3,0